missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals Product Code.: 10153010

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 10153010
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10153010

Brand: Thermo Scientific Acros 153195000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-34-3
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007253
InChI Key VYWYYJYRVSBHJQ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acidShow MoreShow Less
PubChem CID 7433
ChEBI CHEBI:73914
IUPAC Name 3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Density 1.6830g/mL
Assay Percent Range 98% min. (HPLC)
Packaging Plastic bottle
Linear Formula (O2N)2C6H3CO2H
Quantity 500 g
Beilstein 09,413
Merck Index 15,3302
Specific Gravity 1.683
Solubility Information Solubility in water: 1350mg/l (25°C). Other solubilities: very soluble in alcohol and glacial acetic acid,sparingly soluble in ether,benzene and,carbon disulfide
Formula Weight 212.12
Percent Purity 98+%
Chemical Name or Material 3, 5-Dinitrobenzoic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0392478 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number153190010,153191000,153195000
Lot NumberA0448167 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number153190010,153191000,153195000
Lot NumberA0414961 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number153190010,153191000,153195000
Lot NumberA0417088 Certificate TypeCertificate of Analysis Date29/08/2022 Catalog Number153190010,153191000,153195000
Lot NumberA0391323 Certificate TypeCertificate of Analysis Date12/06/2020 Catalog Number153190010,153191000,153195000
compliance-icons
Product Identifier
  • 3,5-Dinitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.