missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dinitrobenzyl alcohol, 98%, Thermo Scientific Chemicals Product Code.: 15475417

3,5-Dinitrobenzyl alcohol, 98%, Thermo Scientific Chemicals

Product Code. 15475417
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15475417

Brand: Thermo Scientific Alfa Aesar H32669.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71022-43-0
Molecular Formula C7H6N2O5
Molecular Weight (g/mol) 198.13
MDL Number MFCD00007235
InChI Key GPHYIQCSMDYRGJ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol #
PubChem CID 116978
IUPAC Name (3,5-dinitrophenyl)methanol
SMILES OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 88°C to 91°C
Quantity 100 g
Beilstein 2054388
Formula Weight 198.13
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzyl alcohol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243398 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog NumberH32669.14,H32669.22,H32669.06
Lot Number10162428 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32669.14,H32669.22,H32669.06
Lot Number10225727 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32669.14,H32669.22,H32669.06
Lot Number10220839 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32669.14,H32669.22,H32669.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dinitrobenzyl alcohol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.