Learn More
3-Amino-5-bromo-2-methoxypyridine, 96%, Thermo Scientific Chemicals
886.98 SEK - 13705.00 SEK
Chemical Identifiers
CAS | 884495-39-0 |
---|---|
Molecular Formula | C6H7BrN2O |
Molecular Weight (g/mol) | 203.039 |
MDL Number | MFCD07368887 |
InChI Key | HJOOFLFWIISCAI-UHFFFAOYSA-N |
Synonym | 3-amino-5-bromo-2-methoxypyridine, 5-bromo-2-methoxy-3-pyridinamine, 3-amino-5-bromopyridin-2-yl methyl ether, 5-bromo-2-methoxy-3-aminopyridine, 5-bromo-2-methoxy-3-cyanopyridine, 5-bromo-2-methyloxy-3-pyridinamine, 3-pyridinamine,5-bromo-2-methoxy, 5-bromo-2-methoxy-pyridin-3-ylamine, pubchem17164, acmc-209qtg |
PubChem CID | 44754869 |
IUPAC Name | 5-bromo-2-methoxypyridin-3-amine |
SMILES | COC1=NC=C(C=C1N)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410628
|
Thermo Scientific Alfa Aesar
H54299.MD |
250 mg | N/A | ||||||
15490618
|
Thermo Scientific Alfa Aesar
H54299.03 |
1 g | N/A | ||||||
15400628
|
Thermo Scientific Alfa Aesar
H54299.06 |
5 g |
13705.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
884495-39-0 | |
203.039 | |
HJOOFLFWIISCAI-UHFFFAOYSA-N | |
44754869 | |
COC1=NC=C(C=C1N)Br |
C6H7BrN2O | |
MFCD07368887 | |
3-amino-5-bromo-2-methoxypyridine, 5-bromo-2-methoxy-3-pyridinamine, 3-amino-5-bromopyridin-2-yl methyl ether, 5-bromo-2-methoxy-3-aminopyridine, 5-bromo-2-methoxy-3-cyanopyridine, 5-bromo-2-methyloxy-3-pyridinamine, 3-pyridinamine,5-bromo-2-methoxy, 5-bromo-2-methoxy-pyridin-3-ylamine, pubchem17164, acmc-209qtg | |
5-bromo-2-methoxypyridin-3-amine |
Specifications
884495-39-0 | |
C6H7BrN2O | |
3-amino-5-bromo-2-methoxypyridine, 5-bromo-2-methoxy-3-pyridinamine, 3-amino-5-bromopyridin-2-yl methyl ether, 5-bromo-2-methoxy-3-aminopyridine, 5-bromo-2-methoxy-3-cyanopyridine, 5-bromo-2-methyloxy-3-pyridinamine, 3-pyridinamine,5-bromo-2-methoxy, 5-bromo-2-methoxy-pyridin-3-ylamine, pubchem17164, acmc-209qtg | |
HJOOFLFWIISCAI-UHFFFAOYSA-N | |
5-bromo-2-methoxypyridin-3-amine | |
44754869 | |
96% |
53°C to 55°C | |
MFCD07368887 | |
Slightly soluble in water. | |
COC1=NC=C(C=C1N)Br | |
203.039 | |
203.04 | |
3-Amino-5-bromo-2-methoxypyridine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.