missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-5-isopropyl-1H-pyrazole hydrochloride, Thermo Scientific™ Product Code.: 15427997

3-Amino-5-isopropyl-1H-pyrazole hydrochloride, Thermo Scientific™

Product Code. 15427997
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15427997

Brand: Thermo Scientific Alfa Aesar H51110.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1347814-85-0
Molecular Formula C6H12ClN3
Molecular Weight (g/mol) 161.63
MDL Number MFCD11111023
InChI Key JHGOPXUCCFVTPD-UHFFFAOYSA-N
Synonym 3-amino-5-isopropyl-1h-pyrazole hydrochloride, 5-isopropyl-1h-pyrazol-3-amine hydrochloride
PubChem CID 53400808
IUPAC Name 5-propan-2-yl-1H-pyrazol-3-amine;hydrochloride
SMILES Cl.CC(C)C1=CC(N)=NN1

Specifications

Quantity 1 g
Formula Weight 161.6
Chemical Name or Material 3-Amino-5-isopropyl-1H-pyrazole hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-5-isopropyl-1H-pyrazole hydrochloride, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.