missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chlorothiophene, 97%, Thermo Scientific™

Product Code. 11324236
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Bromo-2-chlorothiophene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40032-73-3
Molecular Formula C4H2BrClS
Molecular Weight (g/mol) 197.474
MDL Number MFCD00043883
InChI Key KSHOQKKCPJELBV-UHFFFAOYSA-N
Synonym 2-chloro-3-bromo thiophene, 2-chloro-3-bromothiophene, 3-bromo-2-chloro-thiophene, thiophene, 3-bromo-2-chloro, pubchem5259, acmc-1apw4, ksc491i4t, bidd:gt0537, 3-bromo-2-chlorothiophene, inchi=1/c4h2brcls/c5-3-1-2-7-4 3 6/h1-2
PubChem CID 2724559
IUPAC Name 3-bromo-2-chlorothiophene
SMILES C1=CSC(=C1Br)Cl

Specifications

Density 1.808
Boiling Point 70°C (9 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.6
Quantity 1 g
UN Number UN2810
Beilstein 1205539
Solubility Information Not miscible or mix with water.
Formula Weight 197.48
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-chlorothiophene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-2-chlorothiophene, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.