Learn More
3-Bromothiophenol, 95%, Thermo Scientific Chemicals
477.88 SEK - 1551.12 SEK
Chemical Identifiers
CAS | 6320-01-0 |
---|---|
Molecular Formula | C6H5BrS |
Molecular Weight (g/mol) | 189.07 |
MDL Number | MFCD00009603 |
InChI Key | HNGQQUDFJDROPY-UHFFFAOYSA-N |
Synonym | 3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, # |
PubChem CID | 80597 |
IUPAC Name | 3-bromobenzenethiol |
SMILES | SC1=CC=CC(Br)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10195890
|
Thermo Scientific Acros
303710010 |
1 g | Glass bottle |
477.88 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10636364
|
Thermo Scientific Acros
303710050 |
5 g | Glass bottle |
1551.12 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6320-01-0 | |
189.07 | |
HNGQQUDFJDROPY-UHFFFAOYSA-N | |
80597 | |
SC1=CC=CC(Br)=C1 |
C6H5BrS | |
MFCD00009603 | |
3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, # | |
3-bromobenzenethiol |
Specifications
6320-01-0 | |
100.0 | |
1.6320g/mL | |
105°C | |
94% min. (GC) | |
BrC6H4SH | |
MFCD00009603 | |
3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, # | |
SC1=CC=CC(Br)=C1 | |
189.07 | |
189.07 | |
Crystalline Powder |
94.0 | |
White to Beige | |
123°C to 124°C (40.0 mmHg) | |
Authentic | |
C6H5BrS | |
1.6320 to 1.634 | |
1.632 | |
HNGQQUDFJDROPY-UHFFFAOYSA-N | |
3-bromobenzenethiol | |
80597 | |
95% | |
3-Bromothiophenol |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 228-664-9
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.