missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methoxypyridine-4-boronic acid, 95%, Thermo Scientific™

739.07 SEK - 9976.00 SEK

Chemical Identifiers

CAS 957060-88-7
Molecular Formula C6H7BClNO3
Molecular Weight (g/mol) 187.39
MDL Number MFCD06798255
InChI Key YQNAXOGPSUVHNU-UHFFFAOYSA-N
Synonym 3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl
PubChem CID 44119809
IUPAC Name (3-chloro-2-methoxypyridin-4-yl)boronic acid
SMILES COC1=NC=CC(B(O)O)=C1Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
13488618
View Documents
Thermo Scientific Acros
448772500
250 mg Glass bottle
739.07 SEK
250mg
Please to purchase this item. Need a web account? Register with us today!
13468618
View Documents
Thermo Scientific Acros
448770010
1 g Glass bottle
2087.16 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
13478618
View Documents
Thermo Scientific Acros
448770050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

957060-88-7
187.39
YQNAXOGPSUVHNU-UHFFFAOYSA-N
44119809
COC1=NC=CC(B(O)O)=C1Cl
C6H7BClNO3
MFCD06798255
3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl
(3-chloro-2-methoxypyridin-4-yl)boronic acid

Specifications

957060-88-7
94% min. (HPLC)
MFCD06798255
YQNAXOGPSUVHNU-UHFFFAOYSA-N
(3-chloro-2-methoxypyridin-4-yl)boronic acid
44119809
95%
140°C
C6H7BClNO3
3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl
COC1=NC=CC(B(O)O)=C1Cl
187.39
187.39
3-Chloro-2-methoxypyridine-4-boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

Product Identifier
  • 3-Chloro-2-methoxypyridine-4-boronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-2-methoxypyridine-4-boronic acid, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.