Learn More
3-Chloro-2-nitropyridine, 97%, Thermo Scientific Chemicals
Product Code. 11399865
Click to view available options
Quantity:
250 mg
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54231-32-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD00490110
InChI Key PSGASDJUCYTRAD-UHFFFAOYSA-N
Synonym 3-chloro-2-nitro-pyridine, pyridine, 3-chloro-2-nitro, chloronitropyridine, chloro-nitropyridine, pubchem11172, acmc-209lej, 2-nitro-3-chloropyridine, 3-chloro-2-nitro pyridine, #
PubChem CID 554988
IUPAC Name 3-chloro-2-nitropyridine
SMILES C1=CC(=C(N=C1)[N+](=O)[O-])Cl
Specifications
Melting Point 89°C to 92°C
Quantity 1 g
Beilstein 1637069
Formula Weight 158.54
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-nitropyridine