missing translation for 'onlineSavingsMsg'
Learn More
3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 10539882

3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10539882
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10539882

Brand: Thermo Scientific Acros 356480010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2888-06-4
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.07
MDL Number MFCD00051697
InChI Key OINWZUJVEXUHCC-UHFFFAOYSA-N
Synonym 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4bShow MoreShow Less
PubChem CID 17909
IUPAC Name 3-chlorobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.4990g/mL
Boiling Point 102°C (0.9 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4SO2Cl
Refractive Index 1.5670 to 1.569
Quantity 1 g
Specific Gravity 1.499
Formula Weight 211.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-chlorobenzenesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0387656 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog Number356480050,356480250,356480010
Lot NumberA0242567 Certificate TypeCertificate of Analysis Date10/06/2021 Catalog Number356480050,356480250,356480010
compliance-icons
Product Identifier
  • 3-Chlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.