Learn More
3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%, Thermo Scientific Chemicals
288.52 SEK - 2895.00 SEK
Chemical Identifiers
CAS | 769-28-8 |
---|---|
Molecular Formula | C8H8N2O |
Molecular Weight (g/mol) | 148.165 |
MDL Number | MFCD00006269 |
InChI Key | OCYMJCILWYHKAU-UHFFFAOYSA-N |
Synonym | 2-hydroxy-4,6-dimethylnicotinonitrile, 3-cyano-4,6-dimethyl-2-hydroxypyridine, 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 2-hydroxy-4,6-dimethylpyridine-3-carbonitrile, unii-ots4s2r79e, 4,6-dimethyl-2-hydroxynicotinonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, 3-cyano-2-hydroxy-4,6-dimethylpyridine, nicotinonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, ots4s2r79eShow More |
PubChem CID | 69856 |
IUPAC Name | 4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile |
SMILES | CC1=CC(=C(C(=O)N1)C#N)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439713
|
Thermo Scientific Alfa Aesar
A12466.06 |
5 g | N/A | ||||||
11449713
|
Thermo Scientific Alfa Aesar
A12466.14 |
25 g |
940.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454064
|
Thermo Scientific Alfa Aesar
A12466.22 |
100 g |
2895.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
769-28-8 | |
148.165 | |
OCYMJCILWYHKAU-UHFFFAOYSA-N | |
69856 | |
CC1=CC(=C(C(=O)N1)C#N)C |
C8H8N2O | |
MFCD00006269 | |
2-hydroxy-4,6-dimethylnicotinonitrile, 3-cyano-4,6-dimethyl-2-hydroxypyridine, 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 2-hydroxy-4,6-dimethylpyridine-3-carbonitrile, unii-ots4s2r79e, 4,6-dimethyl-2-hydroxynicotinonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, 3-cyano-2-hydroxy-4,6-dimethylpyridine, nicotinonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, ots4s2r79eShow More | |
4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile |
Specifications
769-28-8 | |
C8H8N2O | |
130992 | |
OCYMJCILWYHKAU-UHFFFAOYSA-N | |
4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile | |
69856 | |
98% |
∼290°C (decomposition) | |
MFCD00006269 | |
2-hydroxy-4,6-dimethylnicotinonitrile, 3-cyano-4,6-dimethyl-2-hydroxypyridine, 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 2-hydroxy-4,6-dimethylpyridine-3-carbonitrile, unii-ots4s2r79e, 4,6-dimethyl-2-hydroxynicotinonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, 3-cyano-2-hydroxy-4,6-dimethylpyridine, nicotinonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, ots4s2r79e | |
CC1=CC(=C(C(=O)N1)C#N)C | |
148.165 | |
148.17 | |
3-Cyano-2-hydroxy-4,6-dimethylpyridine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
missing translation for 'einecsNumber' : 212-207-5
missing translation for 'rtecsNumber' : QT3046500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.