Learn More
3-(Di-tert-butylphosphino)propylamine, 10% w/w soln. in THF, Thermo Scientific™
275.00 SEK - 3664.00 SEK
Chemical Identifiers
CAS | 1196147-72-4 |
---|---|
Molecular Formula | C11H26NP |
Molecular Weight (g/mol) | 203.31 |
MDL Number | MFCD17013990 |
InChI Key | PREMCOLELPAPBB-UHFFFAOYSA-N |
Synonym | 3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane |
PubChem CID | 72376443 |
IUPAC Name | 3-ditert-butylphosphanylpropan-1-amine |
SMILES | CC(C)(C)P(CCCN)C(C)(C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1196147-72-4 | |
203.31 | |
PREMCOLELPAPBB-UHFFFAOYSA-N | |
72376443 | |
CC(C)(C)P(CCCN)C(C)(C)C |
C11H26NP | |
MFCD17013990 | |
3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane | |
3-ditert-butylphosphanylpropan-1-amine |
Specifications
1196147-72-4 | |
[(CH3)3C]2PCH2CH2CH2NH2 | |
UN2733 | |
3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane | |
CC(C)(C)P(CCCN)C(C)(C)C | |
203.31 | |
203.3 | |
3-(Di-tert-butylphosphino)propylamine |
C11H26NP | |
MFCD17013990 | |
Air Sensitive | |
PREMCOLELPAPBB-UHFFFAOYSA-N | |
3-ditert-butylphosphanylpropan-1-amine | |
72376443 | |
10% w/w soln. in THF |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335-H336-H351
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon