missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-hexanol, 98%, Thermo Scientific Chemicals

Product Code. 11405816
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-76-2
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00039616
InChI Key WNDLTOTUHMHNOC-UHFFFAOYSA-N
Synonym 3-ethyl-3-hexanol, 3-hexanol, 3-ethyl, 1,1-diethyl-1-butanol, diethyl n-propyl carbinol, acmc-1ak6h
PubChem CID 69008
IUPAC Name 3-ethylhexan-3-ol
SMILES CCCC(O)(CC)CC

Specifications

Density 0.83
Quantity 1 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3-Ethyl-3-hexanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'