missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Product Code. 15494317
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1547-29-1
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD03095042
InChI Key CISUDJRBCKKAGX-UHFFFAOYSA-N
Synonym 3-fluoro-2-hydroxypyridine, 3-fluoropyridin-2-ol, 3-fluoro-2-pyridinol, 2 1h-pyridinone, 3-fluoro, 3-fluoropyridin-2 1h-one, 3-fluoro-pyridin-2-ol, 3-fluoro-2-pyridone, 3-fluoro-2-hydroxypyridine, 3-fluoro-2-pyridone, fluoropyridone, pubchem1200
PubChem CID 2783363
IUPAC Name 3-fluoro-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1)F

Specifications

Melting Point 157°C to 161°C
Odor Odorless
Quantity 5 g
Formula Weight 113.09
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'