Learn More
3-Hydroxypyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
428.00 SEK - 1410.00 SEK
Chemical Identifiers
CAS | 10128-71-9 |
---|---|
Molecular Formula | C6H5NO3 |
Molecular Weight (g/mol) | 139.11 |
MDL Number | MFCD00234165 |
InChI Key | JEHGATQUCUYHJL-UHFFFAOYSA-N |
Synonym | 3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid |
PubChem CID | 459503 |
IUPAC Name | 3-hydroxypyridine-4-carboxylic acid |
SMILES | C1=CN=CC(=C1C(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440179
|
Thermo Scientific Alfa Aesar
H63465.03 |
1 g |
428.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15450179
|
Thermo Scientific Alfa Aesar
H63465.06 |
5 g |
1410.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
10128-71-9 | |
139.11 | |
JEHGATQUCUYHJL-UHFFFAOYSA-N | |
459503 | |
C1=CN=CC(=C1C(=O)O)O |
C6H5NO3 | |
MFCD00234165 | |
3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid | |
3-hydroxypyridine-4-carboxylic acid |
Specifications
10128-71-9 | |
MFCD00234165 | |
3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid | |
JEHGATQUCUYHJL-UHFFFAOYSA-N | |
3-hydroxypyridine-4-carboxylic acid | |
459503 | |
98% | |
3-Hydroxypyridine-4-carboxylic acid |
C6H5NO3 | |
383677 | |
Soluble in methanol. Slightly soluble in water. | |
C1=CN=CC(=C1C(=O)O)O | |
139.11 | |
139.11 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only