Learn More
3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 96-27-5, 7732-18-5 |
---|---|
Molecular Formula | C3H8O2S |
Molecular Weight (g/mol) | 108.16 |
MDL Number | MFCD00004879 |
InChI Key | PJUIMOJAAPLTRJ-UHFFFAOYNA-N |
Synonym | 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol |
PubChem CID | 7291 |
ChEBI | CHEBI:74537 |
IUPAC Name | 3-sulfanylpropane-1,2-diol |
SMILES | OCC(O)CS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10204890
|
Thermo Scientific Acros
125520250 |
25 mL | Glass bottle |
N/A
|
N/A | ||||
10511181
|
Thermo Scientific Acros
125521000 |
100 mL | Glass bottle |
N/A
|
N/A | ||||
13408338
|
Thermo Scientific Acros
125525000 |
500 mL | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-27-5, 7732-18-5 | |
108.16 | |
PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
7291 | |
3-sulfanylpropane-1,2-diol |
C3H8O2S | |
MFCD00004879 | |
3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
CHEBI:74537 | |
OCC(O)CS |
Specifications
96-27-5, 7732-18-5 | |
92.0, 12.0 | |
1.2300g/mL | |
>112°C | |
C3H8O2S | |
1.5073 | |
01,519 | |
1.23 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,insoluble in ether | |
OCC(O)CS | |
108.16 | |
CHEBI:74537 | |
≥88% | |
3-Mercapto-1, 2-propanediol |
88.0, 8.0 | |
Colorless to Yellow | |
118.0°C (5.0 mmHg) | |
Authentic | |
HSCH2CH(OH)CH2OH | |
MFCD00004879 | |
15,9488 | |
3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
3-sulfanylpropane-1,2-diol | |
7291 | |
108.16 | |
Viscous Solution |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-495-0
missing translation for 'rtecsNumber' : TY8140000
missing translation for 'tsca' : TSCA
RUO – Research Use Only