missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxycarbonyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

794.00 SEK - 2890.00 SEK

Chemical Identifiers

CAS 41221-47-0
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00037077
InChI Key WBGWGERFPSYHDT-UHFFFAOYSA-N
Synonym 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate
PubChem CID 2733366
IUPAC Name methyl 3-isocyanatobenzoate
SMILES COC(=O)C1=CC=CC(=C1)N=C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339485
View Documents
Thermo Scientific Alfa Aesar
L18709.03
1 g
794.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11349485
View Documents
Thermo Scientific Alfa Aesar
L18709.06
5 g
2890.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41221-47-0
177.16
WBGWGERFPSYHDT-UHFFFAOYSA-N
2733366
COC(=O)C1=CC=CC(=C1)N=C=O
C9H7NO3
MFCD00037077
3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate
methyl 3-isocyanatobenzoate

Specifications

41221-47-0
>110°C (230°F)
MFCD00037077
Moisture sensitive
WBGWGERFPSYHDT-UHFFFAOYSA-N
methyl 3-isocyanatobenzoate
2733366
97%
35°C to 38°C
C9H7NO3
UN2811
3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate
COC(=O)C1=CC=CC(=C1)N=C=O
177.16
177.16
3-(Methoxycarbonyl)phenyl isocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c

H302+H312+H332-H315-H317-H319-H334-H335

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.