missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals

804.00 SEK - 6910.00 SEK

Chemical Identifiers

CAS 38894-11-0
Molecular Formula C8H12ClN3OS
Molecular Weight (g/mol) 233.714
MDL Number MFCD00149370
InChI Key IYXXQOGEFHAQGU-PIHABLKOSA-N
Synonym 2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochlorideShow MoreShow Less
PubChem CID 6508671
IUPAC Name (E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472436
SDS View Documents
Thermo Scientific Alfa Aesar
H56061.06
5 g
804.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
15482436
SDS View Documents
Thermo Scientific Alfa Aesar
H56061.18
50 g
6910.00 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.