missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-quinoxalinol, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
Specifications
Specifications
CAS | 14003-34-0 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 246°C to 248°C |
Color | Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Plastic bottle |
Molecular Formula | C9H8N2O |
MDL Number | MFCD00006724 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'