missing translation for 'onlineSavingsMsg'
Learn More
3-Nitrobenzamide, 98%, Thermo Scientific Chemicals Product Code.: 11452857

3-Nitrobenzamide, 98%, Thermo Scientific Chemicals

Product Code. 11452857
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11452857

Brand: Thermo Scientific Alfa Aesar A15481.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-09-0
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007984
InChI Key KWAYEPXDGHYGRW-UHFFFAOYSA-N
Synonym m-nitrobenzamide, benzamide, 3-nitro, benzamide, m-nitro, 3-nitro-benzamide, 3-nitrophenylcarboxamide, acmc-209nmr, 3-nitrobenzamide, dsstox_cid_5732, dsstox_rid_77897, dsstox_gsid_25732Show MoreShow Less
PubChem CID 12576
IUPAC Name 3-nitrobenzamide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N

Specifications

Melting Point 140°C to 143°C
Quantity 250 g
Beilstein 777185
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzamide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252067 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberA15481.18,A15481.09,A15481.30
Lot Number10248929 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog NumberA15481.18,A15481.09,A15481.30
Lot Number10214622 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog NumberA15481.18,A15481.09,A15481,A15481.30
Lot Number10214622 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15481.18,A15481.09,A15481.30
Lot NumberF2727A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15481.18,A15481.09,A15481.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Nitrobenzamide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.