missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
374.09 SEK - 1089.21 SEK
Chemical Identifiers
CAS | 85-38-1 |
---|---|
Molecular Formula | C7H5NO5 |
Molecular Weight (g/mol) | 183.12 |
InChI Key | WWWFHFGUOIQNJC-UHFFFAOYSA-N |
Synonym | 3-nitrosalicylic acid, 2-hydroxy-3-nitrobenzoic acid, benzoic acid, 2-hydroxy-3-nitro, salicylic acid, 3-nitro, 2-hydroxy-3-nitro-benzoic acid, 3-nitro-salicylsaure, 3-nitrosalicyclic acid, wln: wnr bq cvq, acmc-209q5qShow More |
PubChem CID | 6807 |
SMILES | C1=CC(=C(C(=C1)[N+](=O)[O-])O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10112874
|
Thermo Scientific Acros
399820010 |
1 g |
374.09 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10431674
|
Thermo Scientific Acros
399820050 |
5 g |
1089.21 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85-38-1 | |
183.12 | |
3-nitrosalicylic acid, 2-hydroxy-3-nitrobenzoic acid, benzoic acid, 2-hydroxy-3-nitro, salicylic acid, 3-nitro, 2-hydroxy-3-nitro-benzoic acid, 3-nitro-salicylsaure, 3-nitrosalicyclic acid, wln: wnr bq cvq, acmc-209q5qShow More | |
C1=CC(=C(C(=C1)[N+](=O)[O-])O)C(=O)O |
C7H5NO5 | |
WWWFHFGUOIQNJC-UHFFFAOYSA-N | |
6807 |
Specifications
85-38-1 | |
97.5% min. (HPLC) | |
C7H5NO5 | |
15,6717 | |
WWWFHFGUOIQNJC-UHFFFAOYSA-N | |
183.12 | |
183.12 | |
3-Nitrosalicylic acid |
142°C to 147°C | |
Glass bottle | |
O2NC6H3(OH)COOH | |
3-nitrosalicylic acid, 2-hydroxy-3-nitrobenzoic acid, benzoic acid, 2-hydroxy-3-nitro, salicylic acid, 3-nitro, 2-hydroxy-3-nitro-benzoic acid, 3-nitro-salicylsaure, 3-nitrosalicyclic acid, wln: wnr bq cvq, acmc-209q5q | |
C1=CC(=C(C(=C1)[N+](=O)[O-])O)C(=O)O | |
6807 | |
98% |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Nitrosalicylic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals