Learn More
3-Pentanol, 98+%, Thermo Scientific Chemicals
374.00 SEK - 2985.00 SEK
Chemical Identifiers
CAS | 584-02-1 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00004574 |
InChI Key | AQIXEPGDORPWBJ-UHFFFAOYSA-N |
Synonym | 3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary |
PubChem CID | 11428 |
ChEBI | CHEBI:77519 |
IUPAC Name | pentan-3-ol |
SMILES | CCC(CC)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342346
|
Thermo Scientific Alfa Aesar
L06065.14 |
25 g |
374.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11352346
|
Thermo Scientific Alfa Aesar
L06065.22 |
100 g |
850.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11362346
|
Thermo Scientific Alfa Aesar
L06065.36 |
500 g |
2985.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.
Solubility
Soluble in acetone, benzene, ethanol and diethyl ether.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
584-02-1 | |
88.15 | |
AQIXEPGDORPWBJ-UHFFFAOYSA-N | |
11428 | |
pentan-3-ol |
C5H12O | |
MFCD00004574 | |
3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary | |
CHEBI:77519 | |
CCC(CC)O |
Specifications
584-02-1 | |
114°C to 115°C | |
Characteristic | |
MFCD00004574 | |
1730964 | |
3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary | |
CCC(CC)O | |
88.15 | |
CHEBI:77519 | |
≥98% |
-8°C | |
33°C (91°F) | |
C5H12O | |
UN1105 | |
14,7120 | |
AQIXEPGDORPWBJ-UHFFFAOYSA-N | |
pentan-3-ol | |
11428 | |
88.15 | |
3-Pentanol |
Safety and Handling
GHS H Statement
H226-H332-H315-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
missing translation for 'rtecsNumber' : SA5075000
missing translation for 'tsca' : TSCA
- 3-Pentanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : 3-Pentanol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.