missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propyl acetate, 98%, Thermo Scientific Chemicals

Product Code. 11439906
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-72-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00026216
InChI Key JRJGKUTZNBZHNK-UHFFFAOYSA-N
Synonym hydrocinnamyl acetate, benzenepropanol, acetate, phenylpropyl acetate, 3-phenyl-1-propyl acetate, 3-acetoxypropyl benzene, 3-phenyl-1-propanol, acetate, 3-acetoxy-1-phenylpropane, 1-acetoxy-3-phenylpropane, benzenepropanol, 1-acetate, benzenepropyl acetate
PubChem CID 31226
ChEBI CHEBI:81257
IUPAC Name 3-phenylpropyl acetate
SMILES CC(=O)OCCCC1=CC=CC=C1

Specifications

Density 1.01
Boiling Point 252°C
Flash Point 130°C (266°F)
Refractive Index 1.496
Quantity 50 g
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material 3-Phenyl-1-propyl acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'