missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylamino)cyclohex-2-ene-1-one, 98%, Thermo Scientific™

Product Code. 10557763
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24706-50-1
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.24
MDL Number MFCD00082654
InChI Key IVONJXTZXFTWNA-UHFFFAOYSA-N
PubChem CID 98795
IUPAC Name 3-anilinocyclohex-2-en-1-one
SMILES O=C1CCCC(NC2=CC=CC=C2)=C1

Specifications

Melting Point 178°C to 182°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 3-anilino-2-cyclohexen-1-one, 2-cyclohexen-1-one, 3-phenylamino, 3-phenylamino cyclohex-2-en-1-one, 3-phenylamino cyclohex-2-enone, 3-anilinocyclohex-ene-1-one, 3-phenylamino-2-cyclohexen-1-one, 2-cyclohexen-1-one,3-phenylamino, acmc-1cr8y, 3-anilino-2-cyclohexenone, aronis25001
Solubility Information Solubility in water: insoluble
Formula Weight 187.24
Percent Purity 98%
Chemical Name or Material 3-(Phenylamino)cyclohex-2-ene-1-one
Product Identifier
  • 3-(Phenylamino)cyclohex-2-ene-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'