missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Phenyllactic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11372297
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 828-01-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00065928
InChI Key VOXXWSYKYCBWHO-UHFFFAOYSA-N
Synonym dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
PubChem CID 3848
ChEBI CHEBI:25998
IUPAC Name 2-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)O

Specifications

Melting Point 95°C to 98°C
Odor Odorless
Quantity 1 g
Beilstein 2209791
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (+/-)-3-Phenyllactic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.