missing translation for 'onlineSavingsMsg'
Learn More

3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals

1722.20 SEK - 5895.00 SEK

Chemical Identifiers

CAS 775351-30-9
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
MDL Number MFCD11044436
InChI Key XMJKBDSKRITXBW-UHFFFAOYSA-N
Synonym 3-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, boronic acid,b-3-2h-tetrazol-5-yl phenyl, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, acmc-209pal
PubChem CID 46737996
IUPAC Name [3-(2H-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC=C1)C1=NNN=N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11916761
View Documents
Thermo Scientific Acros
445560010
1 g
1722.20 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11926761
View Documents
Thermo Scientific Acros
445560050
5 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.