missing translation for 'onlineSavingsMsg'
Learn More

3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one, 97%, Thermo Scientific™

2236.58 SEK - 6857.00 SEK

Chemical Identifiers

CAS 34932-07-5
Molecular Formula C22H19O2P
Molecular Weight (g/mol) 346.37
MDL Number MFCD00065318
InChI Key JUKRJGQYUNADDD-UHFFFAOYSA-N
Synonym 2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
PubChem CID 97111
IUPAC Name 3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
SMILES O=C1OCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11976021
View Documents
Thermo Scientific Acros
440990010
1 g
2236.58 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11986021
View Documents
Thermo Scientific Acros
440990050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

34932-07-5
346.37
JUKRJGQYUNADDD-UHFFFAOYSA-N
97111
O=C1OCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C22H19O2P
MFCD00065318
2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one

Specifications

34932-07-5
100.0
White
Authentic
Glass bottle
MFCD00065318
JUKRJGQYUNADDD-UHFFFAOYSA-N
3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
97111
97%
3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one
96.0
243.0°C to 246.0°C
73.24 to 79.34% (C)
97%
C22H19O2P
2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
O=C1OCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
346.37
346.37
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only