missing translation for 'onlineSavingsMsg'
Learn More
3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific™ Product Code.: 15435109

3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific™

Product Code. 15435109
100mg
Click to view available options
Quantity:
100mg
This item is not returnable. View return policy

Product Code. 15435109

Brand: Thermo Scientific Alfa Aesar J62807.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A selective 5-HT3 serotonin receptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17H21ClN2O2
Molecular Weight (g/mol) 320.817
MDL Number MFCD00210221
InChI Key XIEGSJAEZIGKSA-ICHLHHFLSA-N
Synonym sdz-ics 930
PubChem CID 71300439
IUPAC Name [(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride
SMILES CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl

Specifications

Quantity 100mg
UN Number UN2811
Merck Index 14,9783
Solubility Information Soluble in water
Formula Weight 320.82
Chemical Name or Material 3-Tropanylindole-3-carboxylate hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY01H105 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ62807.ME,J62807.MC
Lot NumberQ09E511 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62807.ME,J62807.MC
Lot NumberQ09E510 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ62807.ME,J62807.MC
Lot NumberF30X086 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ62807.ME,J62807.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.