Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
3880.00 SEK
Specifications
Material | Nickel-plated Brass (Hub), Stainless Steel (304) |
---|---|
Needle Point Style | 5 or AS |
For Use With (Equipment) | TLL Syringes and LT or TLL Connectors |
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
11583254
|
Hamilton™
7751-08 |
17 G |
3880.00 SEK
Pack of 6 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11738386
|
Hamilton™
7751-10 |
19 G |
3880.00 SEK
Pack of 6 |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Nickel-plated Brass (Hub), Stainless Steel (304) | |
5 or AS | |
TLL Syringes and LT or TLL Connectors |