Learn More
Masterflex™ Y-Connector, Barbed Fitting, 316 Stainless Steel
Cole-Parmer™ Y-Connector, Barbed Fitting, 316 Stainless Steel is economical, easy to use, and suitable for low-pressure applications.
Brand: Masterflex™ 31209-55
Additional Details : Weight : 0.01000kg
Description
- Stainless steel construction
- Ideal for low-pressure applications (generally less than 30psi)
- Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Specifications
Autoclavable | |
1/16 in. ID tubing | |
1 Each | |
1/16 in. orifice; 1-1/16 x 9/16 in. | |
Y |
Stainless Steel | |
Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. | |
Fits 1/16 in. ID tubing | |
Autoclave, Ethylene Oxide or Gamma |