Learn More
Nalgene 32 oz. Narrow Mouth Bottle
Click to view available options
Color:
Blue
Gray
Seafoam
Trout Green
Description
- Specially designed lip ideal for keeping the flow, without the stress of a mess
- Tough and guaranteed for life
Specifications
Specifications
Capacity (Metric) | 946 mL |
Capacity (English) | 32 oz. |
Color | Seafoam |
Mouth | Narrow |