Learn More
Greiner Bio-One 384-well, Cell Repellent-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand:  Greiner Bio-One 787979
Product Code. 15712567
  • 18770.00 SEK / Pack of 32
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Description
  • Clear polystyrene construction
  • Solid U-bottom (flat)
  • Cell repellent surface treatment
  • High profile lid
  • Free of heavy metal
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Contents non-cytotoxic
  • For single use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Single-use
384
Cell Repellent
Yes
With Lid
Clear
Polystyrene
Sterile
Round Bottom (U-well)
32/Cs.