Learn More
Memmert™ 4-20 mA Current Loop Interface for UF110plus, Actual Value
Temperature Output for external temperature monitoring.
Brand: Memmert™ UF110plusV3
This item is not returnable.
View return policy
Description
- Temperature controller actual value
Specifications
UF110plus Oven | |
Current Loop Interface | |
4 to 20 mA |
Product Content Correction
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF110plus, Actual Value > For Use With: UF110plus
Spot an opportunity for improvement?Share a Content Correction