Learn More
Memmert™ 4-20 mA Current Loop Interface for UF110plus
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF110plusV6
Product Code. 17528995
  • 5190.00 SEK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UF110plus Oven
Current Loop Interface
4 to 20 mA
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF110plus > For Use With: UF110plus