Learn More
Memmert™ 4-20 mA Current Loop Interface for UF160plus
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF160plusV6
Product Code. 17519105
  • 5190.00 SEK / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UF160plus Oven
Current Loop Interface
4 to 20 mA
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF160plus > For Use With: UF160plus