missing translation for 'onlineSavingsMsg'
Learn More
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™ Product Code.: 10216752

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™

Product Code. 10216752
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10216752

Brand: Thermo Scientific Maybridge CC33839DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-95-3
Molecular Formula C16H24BNO3
Molecular Weight (g/mol) 289.18
MDL Number MFCD03412097
InChI Key NCJDKFFODGZRRL-UHFFFAOYSA-N
Synonym 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacolShow MoreShow Less
PubChem CID 4192663
IUPAC Name 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1CCOCC1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 289.18
Percent Purity 90%
Chemical Name or Material 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.