missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyloctane, 99%, Thermo Scientific Chemicals

435.09 SEK - 1875.00 SEK

Chemical Identifiers

CAS 15869-95-1
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00048813
InChI Key ZMEDGZAGMLTROM-UHFFFAOYSA-N
Synonym 4,4-dimethyl octane, octane, 4,4-dimethyl, acmc-1bqkk
PubChem CID 85151
IUPAC Name 4,4-dimethyloctane
SMILES CCCCC(C)(C)CCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11497134
SDS View Documents
Thermo Scientific Alfa Aesar
B21554.03
1 g N/A N/A N/A
11407144
SDS View Documents
Thermo Scientific Alfa Aesar
B21554.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,4-Dimethyloctane, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.