missing translation for 'onlineSavingsMsg'
Learn More
4,4'-Dipyridyl, 98%, Thermo Scientific Chemicals Product Code.: 10790561

4,4'-Dipyridyl, 98%, Thermo Scientific Chemicals

Product Code. 10790561
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10790561

Brand: Thermo Scientific Acros 276200250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-26-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006416
InChI Key MWVTWFVJZLCBMC-UHFFFAOYSA-N
Synonym 4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cbShow MoreShow Less
PubChem CID 11107
ChEBI CHEBI:30985
IUPAC Name 4-pyridin-4-ylpyridine
SMILES C1=CC(=CC=N1)C1=CC=NC=C1

Specifications

Melting Point 110.0°C to 114.0°C
Color White to Beige
Boiling Point 305.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)-(C5H4N)
Quantity 25 g
Beilstein 23,2
Fieser 16,25
Merck Index 15,3389
Solubility Information Solubility in water: 4,5g/L (25°C). Other solubilities: freely soluble in alcohol, benzene, chloroform,, ether,
Formula Weight 156.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dipyridyl
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462649 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog Number276200250,276201000,276200050
Lot NumberA0439774 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number276200250,276201000,276200050
Lot NumberA0451212 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number276200250,276201000,276200050
Lot NumberA0416674 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number276200250,276201000,276200050
Lot NumberA0428459 Certificate TypeCertificate of Analysis Date31/03/2021 Catalog Number276200250,276201000,276200050
compliance-icons
Product Identifier
  • 4,4`-Dipyridyl, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,4'-Dipyridyl, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.