missing translation for 'onlineSavingsMsg'
Learn More
4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11446366

4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11446366
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11446366

Brand: Thermo Scientific Alfa Aesar B23142.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25519-78-2
Molecular Formula C12H15ClFNO
Molecular Weight (g/mol) 243.71
MDL Number MFCD00044912
InChI Key GPKDBZQZPNOBGM-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl piperidine hydrochloride, 4-fluorophenyl piperidin-4-yl methanone hydrochloride, 4-fluorophenyl 4-piperidinyl ketone hydrochloride, p-fluorophenyl 4-piperidyl ketone hydrochloride, 4-fluorophenyl 4-piperidyl methanone hydrochloride, 4-fluoro-phenyl-piperidin-4-yl-methanone hydrochloride, 4-4-fluorobenzoyl piperidine hcl, 4-p-fluorobenzoyl piperidine hydrochloride, 4-4'-fluorobenzoyl piperidine hydrochloride, methanone, 4-fluorophenyl-4-piperidinyl-, hydrochloride
PubChem CID 3084438
IUPAC Name (4-fluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES [H+].[Cl-].FC1=CC=C(C=C1)C(=O)C1CCNCC1

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 243.71
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.