Learn More
4-(4-Morpholinyl)aniline, 98+%, Thermo Scientific Chemicals
Product Code. 15443217
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
Solubility
Soluble in chloroform, ethyl acetate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2524-67-6
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00006169
InChI Key PHNDZBFLOPIMSM-UHFFFAOYSA-N
Synonym 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
PubChem CID 75655
IUPAC Name 4-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=C(C=C2)N
Specifications
Melting Point 129°C to 133°C
Quantity 1 g
UN Number UN2811
Beilstein 150957
Solubility Information Soluble in chloroform,ethyl acetate.
Formula Weight 178.24
Percent Purity ≥98%
Chemical Name or Material 4-(4-Morpholinyl)aniline