missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Bromo-2-pyridyloxy)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 15438525
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312100-75-6
Molecular Formula C11H7BrClNO3S
Molecular Weight (g/mol) 348.595
MDL Number MFCD21602384
InChI Key PRPZOWBZYLSCTG-UHFFFAOYSA-N
Synonym 4-5-bromo-2-pyridyloxy benzenesulfonyl chloride, 4-5-bromopyridin-2-yl oxy benzenesulfonyl chloride, 4-5-bromopyridin-2-yl oxy-benzenesulfonyl chloride, 4-5-bromopyridin-2-yl oxy benzene-1-sulfonyl chloride
PubChem CID 73995158
IUPAC Name 4-(5-bromopyridin-2-yl)oxybenzenesulfonyl chloride
SMILES C1=CC(=CC=C1OC2=NC=C(C=C2)Br)S(=O)(=O)Cl

Specifications

Melting Point 96°C to 98°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 348.6
Percent Purity 95%
Chemical Name or Material 4-(5-Bromo-2-pyridyloxy)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'