missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminonicotinic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.
Solubility
Soluble in aqueous acid and base.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.
Solubility
Soluble in aqueous acid and base.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | >300°C |
Quantity | 5 g |
Solubility Information | Soluble in aqueous acid and base. |
Formula Weight | 138.13 |
Percent Purity | 98% |
Chemical Name or Material | 4-Aminonicotinic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction