missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylmorpholine-2-carboxylic acid hydrochloride, 97%, Thermo Scientific™
1199.00 SEK - 3948.00 SEK
Chemical Identifiers
CAS | 135072-15-0 |
---|---|
Molecular Formula | C12H15NO3·ClH |
Molecular Weight (g/mol) | 257.72 |
InChI Key | CEDXMALCJZSQHA-UHFFFAOYSA-N |
Synonym | 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6lShow More |
PubChem CID | 2776352 |
IUPAC Name | 4-benzylmorpholine-2-carboxylic acid;hydrochloride |
SMILES | C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl |
Description
Chemical Identifiers
135072-15-0 | |
257.72 | |
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6lShow More | |
4-benzylmorpholine-2-carboxylic acid;hydrochloride |
C12H15NO3·ClH | |
CEDXMALCJZSQHA-UHFFFAOYSA-N | |
2776352 | |
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl |
Specifications
135072-15-0 | |
53.69 to 58.16% (C) 5.22 to 5.65% (N) | |
97% | |
C12H15NO3·ClH | |
CEDXMALCJZSQHA-UHFFFAOYSA-N | |
4-benzylmorpholine-2-carboxylic acid;hydrochloride | |
2776352 | |
97% |
244.0°C to 248.0°C | |
Authentic | |
Glass Bottle | |
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l | |
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl | |
257.72 | |
257.72 | |
4-Benzylmorpholine-2-carboxylic acid hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Benzylmorpholine-2-carboxylic acid hydrochloride, 98%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Benzylmorpholine-2-carboxylic acid hydrochloride, 97%, Thermo Scientific™