Learn More
4-Bromo-1,1,2-trifluoro-1-butene, 98%, Thermo Scientific Chemicals
2540.00 SEK - 11875.00 SEK
Chemical Identifiers
CAS | 10493-44-4 |
---|---|
Molecular Formula | C4H4BrF3 |
Molecular Weight (g/mol) | 188.975 |
MDL Number | MFCD00039274 |
InChI Key | GQCQMFYIFUDARF-UHFFFAOYSA-N |
Synonym | 4-bromo-1,1,2-trifluoro-1-butene, 4-bromo-1,1,2-trifluorobutene-1, 1-butene, 4-bromo-1,1,2-trifluoro, 1,1,2-trifluoro-4-bromobutene, acmc-20amo8, timtec-bb sbb006604, gqcqmfyifudarf-uhfffaoysa, 4-bromo-1,1,2-trifluorobutene., 3,4,4-trifluoro-3-butenyl bromide, 3,4,4-trifluorobut-3-enyl bromide |
PubChem CID | 66333 |
IUPAC Name | 4-bromo-1,1,2-trifluorobut-1-ene |
SMILES | C(CBr)C(=C(F)F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11393356
|
Thermo Scientific Alfa Aesar
L12766.09 |
10 g |
2540.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303366
|
Thermo Scientific Alfa Aesar
L12766.18 |
50 g |
11875.00 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Chemical Identifiers
10493-44-4 | |
188.975 | |
GQCQMFYIFUDARF-UHFFFAOYSA-N | |
66333 | |
C(CBr)C(=C(F)F)F |
C4H4BrF3 | |
MFCD00039274 | |
4-bromo-1,1,2-trifluoro-1-butene, 4-bromo-1,1,2-trifluorobutene-1, 1-butene, 4-bromo-1,1,2-trifluoro, 1,1,2-trifluoro-4-bromobutene, acmc-20amo8, timtec-bb sbb006604, gqcqmfyifudarf-uhfffaoysa, 4-bromo-1,1,2-trifluorobutene., 3,4,4-trifluoro-3-butenyl bromide, 3,4,4-trifluorobut-3-enyl bromide | |
4-bromo-1,1,2-trifluorobut-1-ene |
Specifications
10493-44-4 | |
95°C to 98°C | |
Recognizable | |
BrCH2CH2CF=CF2 | |
MFCD00039274 | |
1840755 | |
4-bromo-1,1,2-trifluoro-1-butene, 4-bromo-1,1,2-trifluorobutene-1, 1-butene, 4-bromo-1,1,2-trifluoro, 1,1,2-trifluoro-4-bromobutene, acmc-20amo8, timtec-bb sbb006604, gqcqmfyifudarf-uhfffaoysa, 4-bromo-1,1,2-trifluorobutene., 3,4,4-trifluoro-3-butenyl bromide, 3,4,4-trifluorobut-3-enyl bromide | |
GQCQMFYIFUDARF-UHFFFAOYSA-N | |
4-bromo-1,1,2-trifluorobut-1-ene | |
66333 | |
98% |
1.639 | |
17°C (62°F) | |
C4H4BrF3 | |
1.402 | |
UN1993 | |
Light sensitive | |
Not miscible or difficult to mix in water. | |
C(CBr)C(=C(F)F)F | |
188.975 | |
188.98 | |
4-Bromo-1,1,2-trifluoro-1-butene |
Safety and Handling
GHS H Statement
H225-H319-H335
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 234-019-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.