missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11392549
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142808-15-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD01075254
InChI Key OEPBVXQEVBURGC-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzotrifluoride, 4-bromo-2-fluoro-1-trifluoromethyl benzene, 2-fluoro-4-bromobenzotrifluoride, 1-bromo-3-fluoro-4-trifluoromethyl benzene, benzene, 4-bromo-2-fluoro-1-trifluoromethyl, 2-fluoro-4-bromotrifluorotoluene, 3-fluoro-4-trifluoromethyl bromobenzene, 4-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 4-bromo-2-fluoro benzotrifluoride, 4-bromo-2-fluoro-1-trifluoromethyl-benzene
PubChem CID 2782904
IUPAC Name 4-bromo-2-fluoro-1-(trifluoromethyl)benzene
SMILES FC1=C(C=CC(Br)=C1)C(F)(F)F

Specifications

Refractive Index 1.4615
Quantity 5 g
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluorobenzotrifluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'