missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11382016

4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11382016
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11382016

Brand: Thermo Scientific Alfa Aesar L03071.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68837-59-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00040905
InChI Key RVCJOGNLYVNRDN-UHFFFAOYSA-N
Synonym 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5Show MoreShow Less
PubChem CID 99570
IUPAC Name 4-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Br)C(=O)O

Specifications

Melting Point 182°C to 184°C
Quantity 25 g
Beilstein 2249816
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-methylbenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI16T031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot Number10148778 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot Number10125703 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot NumberL01Q074 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot NumberF13R024 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.