missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-bromobenzaldehyde, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Linear Formula | BrC6H4CHO |
Quantity | 50 g |
Formula Weight | 185.02 |
Percent Purity | 95% |
Chemical Name or Material | 4-Bromobenzaldehyde |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- 4-Bromobenzaldehyde
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P264-Wash face, hands an any exposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 4-bromo-benzaldehyde
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'