missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

1975.00 SEK - 6155.00 SEK

Chemical Identifiers

CAS 26177-44-6
Molecular Formula C7H9BrClN
Molecular Weight (g/mol) 222.51
MDL Number MFCD00012860
InChI Key BVYUYDBWQMSOKM-UHFFFAOYSA-N
Synonym 4-bromobenzylamine hydrochloride, 4-bromophenyl methanamine hydrochloride, 4-bromobenzylamine hcl, 4-bba x hcl, 4-bromo-benzylamine hydrochloride, p-bromobenzylamine hydrochloride, benzenemethanamine, 4-bromo-, hydrochloride, pubchem3751, acmc-1cfrz, 4-bromobenzylamine, hcl
PubChem CID 2724096
IUPAC Name (4-bromophenyl)methanamine;hydrochloride
SMILES [H+].[Cl-].NCC1=CC=C(Br)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11464423
SDS View Documents
Thermo Scientific Alfa Aesar
A10979.09
10 g
1975.00 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
11474423
SDS View Documents
Thermo Scientific Alfa Aesar
A10979.18
50 g
6155.00 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Bromobenzylamine hydrochloride is used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine. Further, it is used to prepare 1-(4-Brombenzyl)-pyrrole by reacting with 2,5-dimethoxy-tetrahydro-furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzylamine hydrochloride is used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine. Further, it is used to prepare 1-(4-Brombenzyl)-pyrrole by reacting with 2,5-dimethoxy-tetrahydro-furan.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.