missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15460068
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137504-86-0
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD01319010
InChI Key CMJQIHGBUKZEHP-UHFFFAOYSA-N
Synonym 4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl, 4-chloro-3-fluorobenzeneboronicacid, pubchem1776, ksc174k0r, acmc-1c195, 4-chloro3-fluorophenylboronic acid
PubChem CID 2778656
IUPAC Name (4-chloro-3-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)Cl)F)(O)O

Specifications

Melting Point 198°C to 204°C
Quantity 1 g
Formula Weight 174.37
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluorobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'