Learn More
4-Chloro-3-methylpyridine hydrochloride, 97%, Thermo Scientific Chemicals
620.00 SEK - 1540.00 SEK
Chemical Identifiers
CAS 19524-08-4
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD03092890
InChI Key WPIUSHWGBZMBHZ-UHFFFAOYSA-N
Synonym 4-chloro-3-methylpyridine hydrochloride, 4-chloro-3-picoline hcl, 4-chloro-3-methylpyridine hcl, 4-chloro-3-picoline hydrochloride, pyridine, 4-chloro-3-methyl-, hydrochloride, 4-chloro-3-methylpyridinehydrochloride, acmc-20amlc, pubchem9218, ksc495e6r, 4-chloro-3methylpyridine hydrochlorideShow MoreShow Less
PubChem CID 16217655
IUPAC Name 4-chloro-3-methylpyridine;hydrochloride
SMILES CC1=C(C=CN=C1)Cl.Cl
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355325
SDS View Documents
Thermo Scientific Alfa Aesar
L19558.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11348956
SDS View Documents
Thermo Scientific Alfa Aesar
L19558.03
1 g
1540.00 SEK
1g
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Show More
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
4-Chloro-3-methylpyridine hydrochloride, 97%, Thermo Scientific Chemicals