missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Chloro-3-(trifluoromethoxy)benzoic acid is used as pharmaceuticals intermediates.
Solubility
Sparingly Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Chloro-3-(trifluoromethoxy)benzoic acid is used as pharmaceuticals intermediates.
Solubility
Sparingly Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 123°C to 126°C |
Quantity | 1 g |
Solubility Information | Sparingly Soluble in water. |
Formula Weight | 240.57 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-3-(trifluoromethoxy)benzoic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Chloro-3-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction